missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylphosphonate, 99%, Thermo Scientific™

$141.57 - $1,691.72

Chemical Identifiers

CAS 1080-32-6
Molecular Formula C11H17O3P
Molecular Weight (g/mol) 228.23
InChI Key AIPRAPZUGUTQKX-UHFFFAOYSA-N
Synonym diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
PubChem CID 14122
IUPAC Name diethoxyphosphorylmethylbenzene
SMILES CCOP(=O)(CC1=CC=CC=C1)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC114170250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC114171000
View Documents Promotion Details
N/A 100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC114175000
View Documents Promotion Details
N/A 500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

1080-32-6
228.23
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
diethoxyphosphorylmethylbenzene
C11H17O3P
AIPRAPZUGUTQKX-UHFFFAOYSA-N
14122
CCOP(=O)(CC1=CC=CC=C1)OCC

Specifications

1080-32-6
106.0°C to 108.0°C (1.0 mmHg)
Authentic
C11H17O3P
(C2H5O)2P(O)CH2C6H5
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
CCOP(=O)(CC1=CC=CC=C1)OCC
228.23
228.23
Diethyl benzylphosphonate
1.0900g/mL
>112°C
99%
1.4950 to 1.4980
1.09
AIPRAPZUGUTQKX-UHFFFAOYSA-N
diethoxyphosphorylmethylbenzene
14122
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 214-097-4

SDS
Documents

Documents

RUO – Research Use Only