missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylphosphonate, 99%

Catalog No. AC114175000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114175000 500 g Glass bottle
AC114170250 25 g Glass bottle
AC114171000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114175000 Supplier Thermo Scientific Chemicals Supplier No. 114175000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1080-32-6 | C11H17O3P | 228.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 1080-32-6
Molecular Formula C11H17O3P
Molecular Weight (g/mol) 228.23
InChI Key AIPRAPZUGUTQKX-UHFFFAOYSA-N
Synonym diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
PubChem CID 14122
IUPAC Name diethoxyphosphorylmethylbenzene
SMILES CCOP(=O)(CC1=CC=CC=C1)OCC

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 106.0°C to 108.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4950 to 1.4980
Linear Formula (C2H5O)2P(O)CH2C6H5
Quantity 500 g
Specific Gravity 1.09
Formula Weight 228.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl benzylphosphonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 214-097-4

RUO – Research Use Only