missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylphosphonate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 114171000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC114171000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

1080-32-6
228.23
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
diethoxyphosphorylmethylbenzene
C11H17O3P
AIPRAPZUGUTQKX-UHFFFAOYSA-N
14122
CCOP(=O)(CC1=CC=CC=C1)OCC

Specifications

Diethyl benzylphosphonate
1.0900g/mL
>112°C
99%
C11H17O3P
(C2H5O)2P(O)CH2C6H5
1.09
AIPRAPZUGUTQKX-UHFFFAOYSA-N
diethoxyphosphorylmethylbenzene
14122
99%
1080-32-6
106.0°C to 108.0°C (1.0 mmHg)
Authentic
Glass bottle
1.4950 to 1.4980
100g
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
CCOP(=O)(CC1=CC=CC=C1)OCC
228.23
228.23
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 214-097-4

SDS
Documents

Documents

Promotions

Promotions