missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Iodobenzyl)phosphonate 98.0+%, TCI America™

Catalog No. D368925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D368925G 25 g
D36895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D368925G Supplier TCI America Supplier No. D368925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 173443-43-1
Molecular Formula C11H16IO3P
Molecular Weight (g/mol) 354.124
MDL Number MFCD09033365
InChI Key ACYSYNIYTMHNSY-UHFFFAOYSA-N
Synonym (4-Iodobenzyl)phosphonic Acid Diethyl Ester
PubChem CID 15166405
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-iodobenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)I)OCC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 354.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Iodobenzyl)phosphonate
EINECSNumber (3)-4513
TSCA No