accessibility menu, dialog, popup

UserName

CAS RN 173443-43-1

IUPAC Name: diethyl [(4-iodophenyl)methyl]phosphonate
Synonyms: diethyl (4-iodophenyl)methylphosphonate
Molecular Weight (g/mol): 354.12
Molecular Formula: C11H16IO3P
InChi Key: ACYSYNIYTMHNSY-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(I)C=C1)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results