missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Fluorobenzyl)phosphonate 98.0+%, TCI America™

Catalog No. D33245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33245G 5 g
D332425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33245G Supplier TCI America Supplier No. D33245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63909-58-0
Molecular Formula C11H16FO3P
Molecular Weight (g/mol) 246.22
MDL Number MFCD06795966
InChI Key FIYRZOAUPPNGAO-UHFFFAOYSA-N
Synonym (4-Fluorobenzyl)phosphonic Acid Diethyl Ester
PubChem CID 11064685
IUPAC Name diethyl [(4-fluorophenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC=C(F)C=C1)OCC

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 246.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Fluorobenzyl)phosphonate