accessibility menu, dialog, popup

UserName

CAS RN 63909-58-0

IUPAC Name: diethyl [(4-fluorophenyl)methyl]phosphonate
Synonyms: diethyl (4-fluorophenyl)methylphosphonate
Molecular Weight (g/mol): 246.22
Molecular Formula: C11H16FO3P
InChi Key: FIYRZOAUPPNGAO-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(F)C=C1)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results