missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Bromophenyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7128774
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5453200MG 200 mg
D54531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5453200MG Supplier TCI America Supplier No. D5453200MG

Chemical Identifiers

CAS 20677-12-7
Molecular Formula C10H14BrO3P
Molecular Weight (g/mol) 293.10
MDL Number MFCD00769296
InChI Key WBJRWCXJMRZDPA-UHFFFAOYSA-N
Synonym (4-Bromophenyl)phosphonic Acid Diethyl Ester
PubChem CID 12625835
IUPAC Name diethyl (4-bromophenyl)phosphonate
SMILES CCOP(=O)(OCC)C1=CC=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 120°C
Quantity 200 mg
Formula Weight 293.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Bromophenyl)phosphonate