accessibility menu, dialog, popup

UserName

CH3OPOBrOCH3

CAS RN 20677-12-7

IUPAC Name: diethyl (4-bromophenyl)phosphonate
Synonyms: diethyl 4-bromophenylphosphonate
Molecular Weight (g/mol): 293.1
Molecular Formula: C10H14BrO3P
InChi Key: WBJRWCXJMRZDPA-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results