missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-Bromobenzyl)phosphonate 98.0+%, TCI America™

Catalog No. D36885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36885G 5 g
D368825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36885G Supplier TCI America Supplier No. D36885G

Chemical Identifiers

CAS 38186-51-5
Molecular Formula C11H16BrO3P
Molecular Weight (g/mol) 307.124
MDL Number MFCD05663693
InChI Key IPTXXSZUISGKCJ-UHFFFAOYSA-N
Synonym (4-Bromobenzyl)phosphonic Acid Diethyl Ester
PubChem CID 11077666
IUPAC Name 1-bromo-4-(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)Br)OCC

Specifications

Color Yellow
Boiling Point 183°C
Quantity 5 g
Formula Weight 307.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (4-Bromobenzyl)phosphonate