accessibility menu, dialog, popup

UserName

H3COPOBrOCH3

CAS RN 38186-51-5

IUPAC Name: diethyl [(4-bromophenyl)methyl]phosphonate
Synonyms: diethyl (4-bromophenyl)methylphosphonate
Molecular Weight (g/mol): 307.12
Molecular Formula: C11H16BrO3P
InChi Key: IPTXXSZUISGKCJ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=C(Br)C=C1)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results