missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (3-Methoxybenzyl)phosphonate 98.0+%, TCI America™
$158.44 - $480.92
Chemical Identifiers
| CAS | 60815-18-1 |
|---|---|
| Molecular Formula | C12H19O4P |
| Molecular Weight (g/mol) | 258.25 |
| MDL Number | MFCD08276363 |
| InChI Key | IGGMUQNGGBRXFB-UHFFFAOYSA-N |
| Synonym | 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester |
| PubChem CID | 10611200 |
| IUPAC Name | diethyl [(3-methoxyphenyl)methyl]phosphonate |
| SMILES | CCOP(=O)(CC1=CC(OC)=CC=C1)OCC |
Chemical Identifiers
| 60815-18-1 | |
| 258.25 | |
| IGGMUQNGGBRXFB-UHFFFAOYSA-N | |
| 10611200 | |
| CCOP(=O)(CC1=CC(OC)=CC=C1)OCC |
| C12H19O4P | |
| MFCD08276363 | |
| 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester | |
| diethyl [(3-methoxyphenyl)methyl]phosphonate |
Specifications
| 60815-18-1 | |
| 135°C | |
| MFCD08276363 | |
| 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester | |
| CCOP(=O)(CC1=CC(OC)=CC=C1)OCC | |
| 258.25 | |
| 258.25 | |
| Liquid |
| Yellow | |
| C12H19O4P | |
| 5 g | |
| IGGMUQNGGBRXFB-UHFFFAOYSA-N | |
| diethyl [(3-methoxyphenyl)methyl]phosphonate | |
| 10611200 | |
| ≥98.0% (GC) | |
| Diethyl (3-Methoxybenzyl)phosphonate |
Safety and Handling
TSCA : No