missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (3-Methoxybenzyl)phosphonate 98.0+%, TCI America™
Supplier: TCI America D332625G
Specifications
| Diethyl (3-Methoxybenzyl)phosphonate | |
| Yellow | |
| C12H19O4P | |
| 25 g | |
| IGGMUQNGGBRXFB-UHFFFAOYSA-N | |
| diethyl [(3-methoxyphenyl)methyl]phosphonate | |
| 10611200 | |
| ≥98.0% (GC) |
| 60815-18-1 | |
| 135°C | |
| MFCD08276363 | |
| 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester | |
| CCOP(=O)(CC1=CC(OC)=CC=C1)OCC | |
| 258.25 | |
| 258.25 | |
| Liquid |
Chemical Identifiers
| 60815-18-1 | |
| 258.25 | |
| IGGMUQNGGBRXFB-UHFFFAOYSA-N | |
| 10611200 | |
| CCOP(=O)(CC1=CC(OC)=CC=C1)OCC |
| C12H19O4P | |
| MFCD08276363 | |
| 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester | |
| diethyl [(3-methoxyphenyl)methyl]phosphonate |
Safety and Handling
TSCA : No