CAS RN 60815-18-1
CAS RN 60815-18-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diethyl (3-Methoxybenzyl)phosphonate 98.0+%, TCI America™
CAS: 60815-18-1 Molecular Formula: C12H19O4P Molecular Weight (g/mol): 258.25 MDL Number: MFCD08276363 InChI Key: IGGMUQNGGBRXFB-UHFFFAOYSA-N Synonym: 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester PubChem CID: 10611200 IUPAC Name: diethyl [(3-methoxyphenyl)methyl]phosphonate SMILES: CCOP(=O)(CC1=CC(OC)=CC=C1)OCC