missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 3,5-Dimethyl-2,4-pyrroledicarboxylate 98.0+%, TCI America™
Supplier: TCI America D401225G
Specifications
| Diethyl 3,5-Dimethyl-2,4-pyrroledicarboxylate | |
| 136°C | |
| MFCD00005218 | |
| diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylic acid diethyl ester, knorr's pyrrole, 2,4-diethyl 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylate, 1h-pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, 2,4-dimethyl-3,5-dicarbethoxypyrrole, diethyl 2,4-dimethyl-1h-pyrrole-3,5-dicarboxylate, diethyl2,4-dimethylpyrrole-3,5-dicarboxylate, pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, diethyl 2 ,4-dimethylpyrrole-3,5-dicarboxylate | |
| CCOC(=O)C1=C(NC(=C1C)C(=O)OCC)C | |
| 239.271 | |
| 239.27 | |
| Crystalline Powder |
| 2436-79-5 | |
| C12H17NO4 | |
| 25 g | |
| XSBSXJAYEPDGSF-UHFFFAOYSA-N | |
| diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate | |
| 75526 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2436-79-5 | |
| 239.271 | |
| XSBSXJAYEPDGSF-UHFFFAOYSA-N | |
| diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate |
| C12H17NO4 | |
| MFCD00005218 | |
| 75526 | |
| CCOC(=O)C1=C(NC(=C1C)C(=O)OCC)C |
Safety and Handling
TSCA : No