accessibility menu, dialog, popup

UserName

CH3OONHCH3H3COOCH3

CAS RN 2436-79-5

IUPAC Name: 2,4-diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate
Synonyms: 2,4-diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate
Molecular Weight (g/mol): 239.27
Molecular Formula: C12H17NO4
InChi Key: XSBSXJAYEPDGSF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)C(C(=O)OCC)=C(C)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results