missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3,5-Dimethyl-2,4-pyrroledicarboxylate 98.0+%, TCI America™

Catalog No. p-7128050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40125G 5 g
D401225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40125G Supplier TCI America Supplier No. D40125G
Only null left

Chemical Identifiers

CAS 2436-79-5
Molecular Formula C12H17NO4
Molecular Weight (g/mol) 239.271
MDL Number MFCD00005218
InChI Key XSBSXJAYEPDGSF-UHFFFAOYSA-N
PubChem CID 75526
IUPAC Name diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C1C)C(=O)OCC)C

Specifications

Melting Point 136°C
Quantity 5 g
Synonym diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylic acid diethyl ester, knorr's pyrrole, 2,4-diethyl 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylate, 1h-pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, 2,4-dimethyl-3,5-dicarbethoxypyrrole, diethyl 2,4-dimethyl-1h-pyrrole-3,5-dicarboxylate, diethyl2,4-dimethylpyrrole-3,5-dicarboxylate, pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, diethyl 2 ,4-dimethylpyrrole-3,5-dicarboxylate
Formula Weight 239.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 3,5-Dimethyl-2,4-pyrroledicarboxylate