missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethanolamine 99.0+%, TCI America™
Supplier: TCI America S037610G
Specifications
| Diethanolamine [Matrix for FABMS and liquid SIMS] | |
| White | |
| C4H11NO2 | |
| 10 g | |
| ZBCBWPMODOFKDW-UHFFFAOYSA-N | |
| 2-[(2-hydroxyethyl)amino]ethan-1-ol | |
| 8113 | |
| 105.14 | |
| Crystalline Lumps |
| 111-42-2 | |
| 11.0 (0.1N H2O soln.) | |
| MFCD00002843 | |
| diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis | |
| OCCNCCO | |
| 105.14 | |
| CHEBI:28123 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 111-42-2 | |
| 105.14 | |
| ZBCBWPMODOFKDW-UHFFFAOYSA-N | |
| 8113 | |
| 2-[(2-hydroxyethyl)amino]ethan-1-ol |
| C4H11NO2 | |
| MFCD00002843 | |
| diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis | |
| CHEBI:28123 | |
| OCCNCCO |
Safety and Handling
EINECSNumber : (2)-0302&(2)-0354
RTECSNumber : KL2975000
TSCA : Yes