missing translation for 'onlineSavingsMsg'
Learn More

Diethanolamine 99.0+%, TCI America™

Catalog No. p-7135870
Change view
Click to view available options
Quantity:
1 g
10 g
Catalog No. Quantity
S03761G 1 g
S037610G 10 g
2 options

Catalog No. S03761G

Supplier: TCI America S03761G

Only null left

Chemical Identifiers

CAS 111-42-2
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00002843
InChI Key ZBCBWPMODOFKDW-UHFFFAOYSA-N
Synonym diethanolamine, 2,2'-iminodiethanol, diolamine, iminodiethanol, bis 2-hydroxyethyl amine, diethylolamine, n,n-diethanolamine, diethanolamin, 2,2'-dihydroxydiethylamine, ethanol, 2,2'-iminobis
PubChem CID 8113
ChEBI CHEBI:28123
IUPAC Name 2-[(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCNCCO

Specifications

Color White
pH 11.0 (0.1N H2O soln.)
Quantity 1 g
Formula Weight 105.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Diethanolamine [Matrix for FABMS and liquid SIMS]
EINECSNumber (2)-0302&(2)-0354
RTECSNumber KL2975000
TSCA Yes