missing translation for 'onlineSavingsMsg'
Learn More

Diesel Fuel #2 Composite Standard, Restek™

Catalog No. p-3200467
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06801429 1 mL
06801433 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06801429 Supplier Chromatographic Supplier No. RK 31258
Only null left

Specifications

Chemical Name or Material Diesel Fuel #2 Composite Standard
Packaging Ampule
Solvent or Matrix Methylene Chloride
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 50,000μg/mL