missing translation for 'onlineSavingsMsg'
Learn More

Dienogest 98.0+%, TCI America™

Supplier:  TCI America D523050MG

 View more versions of this product

Catalog No. D523050MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dienogest
215°C
C20H25NO2
50 mg
AZFLJNIPTRTECV-FUMNGEBKSA-N
2-[(8S,13S,14S,17R)-17-hydroxy-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]acetonitrile
68861
311.43
Crystalline Powder
65928-58-7
White
MFCD00868356
17alpha-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Cyanomethyl-17-hydroxy-estra-4,9-dien-3-one
CC12CCC3=C4CCC(=O)C=C4CCC3C1CCC2(CC#N)O
311.425
CHEBI:70708
≥98.0% (HPLC)

Chemical Identifiers

65928-58-7
311.425
AZFLJNIPTRTECV-FUMNGEBKSA-N
68861
2-[(8S,13S,14S,17R)-17-hydroxy-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]acetonitrile
C20H25NO2
MFCD00868356
17alpha-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Cyanomethyl-17-hydroxy-estra-4,9-dien-3-one
CHEBI:70708
CC12CCC3=C4CCC(=O)C=C4CCC3C1CCC2(CC#N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : RC8899500

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions