missing translation for 'onlineSavingsMsg'
Learn More

Dienogest 98.0+%, TCI America™

Catalog No. p-7128698
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D523050MG 50 mg
D5230250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D523050MG Supplier TCI America Supplier No. D523050MG
Only null left

Chemical Identifiers

CAS 65928-58-7
Molecular Formula C20H25NO2
Molecular Weight (g/mol) 311.425
MDL Number MFCD00868356
InChI Key AZFLJNIPTRTECV-FUMNGEBKSA-N
Synonym 17alpha-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Cyanomethyl-17-hydroxy-estra-4,9-dien-3-one
PubChem CID 68861
ChEBI CHEBI:70708
IUPAC Name 2-[(8S,13S,14S,17R)-17-hydroxy-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]acetonitrile
SMILES CC12CCC3=C4CCC(=O)C=C4CCC3C1CCC2(CC#N)O

Specifications

Melting Point 215°C
Color White
Quantity 50 mg
Formula Weight 311.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dienogest
RTECSNumber RC8899500
TSCA No
Recommended Storage Refrigerator