missing translation for 'onlineSavingsMsg'
Learn More

Didodecyl phthalate, 97%

Catalog No. AC407130250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407130250 Supplier Thermo Scientific Chemicals Supplier No. 407130250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2432-90-8 | C32H54O4 | 502.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2432-90-8
Molecular Formula C32H54O4
Molecular Weight (g/mol) 502.77
MDL Number MFCD00041920
InChI Key PUFGCEQWYLJYNJ-UHFFFAOYSA-N
Synonym didodecyl phthalate, dilauryl phthalate, didodecylphthalate, 1,2-benzenedicarboxylic acid, didodecyl ester, di-n-dodecyl phthalate, phthalic acid, didodecyl ester, 1,2-benzenedicarboxylicacid, 1,2-didodecyl ester, 1,2-benzenedicarboxylic acid, 1,2-didodecyl ester, 1, didodecyl ester, phthalic acid bis-dodecyl ester
PubChem CID 17082
IUPAC Name didodecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCCCC

Specifications

Melting Point 21.0°C to 23.0°C
Color Yellow to Tan
Density 0.9400g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.4820
Quantity 25 g
Beilstein 09, IV, 3186
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, diethyl ether, benzene
Formula Weight 502.77
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Didodecyl phthalate, 98%
Show More Show Less
EINECSNumber 219-415-5

RUO – Research Use Only