accessibility menu, dialog, popup

UserName

CAS RN 2432-90-8

IUPAC Name: 1,2-didodecyl benzene-1,2-dicarboxylate
Synonyms: didodecylphthalate
Molecular Weight (g/mol): 502.78
Molecular Formula: C32H54O4
InChi Key: PUFGCEQWYLJYNJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results