missing translation for 'onlineSavingsMsg'
Learn More

Didodecyl 3,3'-Thiodipropionate 90.0+%, TCI America™

$57.75 - $163.64

Chemical Identifiers

CAS 123-28-4
Molecular Formula C30H58O4S
Molecular Weight (g/mol) 514.85
MDL Number MFCD00026589
InChI Key GHKOFFNLGXMVNJ-UHFFFAOYSA-N
Synonym 3,3′C-Thiodipropionic Acid Didodecyl Ester, Dilauryl 3,3′C-Thiodipropionate
PubChem CID 31250
IUPAC Name dodecyl 3-(3-dodecoxy-3-oxopropyl)sulfanylpropanoate
SMILES CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T020525G
View Documents
TCI America
T020525G
25 g
Each for $57.75
Only null left
 
T0205500G
View Documents
TCI America
T0205500G
500 g
Each for $163.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

123-28-4
514.85
GHKOFFNLGXMVNJ-UHFFFAOYSA-N
31250
CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC
C30H58O4S
MFCD00026589
3,3′C-Thiodipropionic Acid Didodecyl Ester, Dilauryl 3,3′C-Thiodipropionate
dodecyl 3-(3-dodecoxy-3-oxopropyl)sulfanylpropanoate

Specifications

123-28-4
White
MFCD00026589
3,3′C-Thiodipropionic Acid Didodecyl Ester, Dilauryl 3,3′C-Thiodipropionate
CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC
514.85
514.85
Crystalline Powder
42°C
C30H58O4S
25 g
GHKOFFNLGXMVNJ-UHFFFAOYSA-N
dodecyl 3-(3-dodecoxy-3-oxopropyl)sulfanylpropanoate
31250
≥90.0% (GC)
Didodecyl 3,3′-Thiodipropionate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0919&(2)-1399

RTECSNumber : UF8000000

TSCA : Yes

SDS
Documents

Documents