missing translation for 'onlineSavingsMsg'
Learn More

Didodecyl 3,3'-Thiodipropionate 90.0+%, TCI America™

Catalog No. T020525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T020525G 25 g
T0205500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T020525G Supplier TCI America Supplier No. T020525G
Only null left

Chemical Identifiers

CAS 123-28-4
Molecular Formula C30H58O4S
Molecular Weight (g/mol) 514.85
MDL Number MFCD00026589
InChI Key GHKOFFNLGXMVNJ-UHFFFAOYSA-N
Synonym 3,3′C-Thiodipropionic Acid Didodecyl Ester, Dilauryl 3,3′C-Thiodipropionate
PubChem CID 31250
IUPAC Name dodecyl 3-(3-dodecoxy-3-oxopropyl)sulfanylpropanoate
SMILES CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC

Specifications

Melting Point 42°C
Color White
Quantity 25 g
Formula Weight 514.85
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Didodecyl 3,3′-Thiodipropionate
EINECSNumber (2)-0919&(2)-1399
RTECSNumber UF8000000
TSCA Yes