missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentyl(dimethoxy)silane 98.0+%, TCI America™

Supplier:  TCI America D439625G

 View more versions of this product

Catalog No. D439625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dicyclopentyl(dimethoxy)silane
Colorless
C12H24O2Si
25 g
JWCYDYZLEAQGJJ-UHFFFAOYSA-N
dicyclopentyldimethoxysilane
9794573
≥98.0% (GC)
126990-35-0
120°C
MFCD01074471
3082
CO[Si](OC)(C1CCCC1)C1CCCC1
228.41
228.41
Liquid

Chemical Identifiers

126990-35-0
228.41
JWCYDYZLEAQGJJ-UHFFFAOYSA-N
dicyclopentyldimethoxysilane
C12H24O2Si
MFCD01074471
9794573
CO[Si](OC)(C1CCCC1)C1CCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4264

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions