accessibility menu, dialog, popup

UserName

H3COSiOCH3

CAS RN 126990-35-0

IUPAC Name: dicyclopentyldimethoxysilane
Synonyms: dicyclopentyldimethoxysilane
Molecular Weight (g/mol): 228.41
Molecular Formula: C12H24O2Si
InChi Key: JWCYDYZLEAQGJJ-UHFFFAOYSA-N
SMILES: CO[Si](OC)(C1CCCC1)C1CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results