missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentyl(dimethoxy)silane 98.0+%, TCI America™

Catalog No. p-7128255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43965G 5 g
D439625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43965G Supplier TCI America Supplier No. D43965G

Chemical Identifiers

CAS 126990-35-0
Molecular Formula C12H24O2Si
Molecular Weight (g/mol) 228.41
MDL Number MFCD01074471
InChI Key JWCYDYZLEAQGJJ-UHFFFAOYSA-N
PubChem CID 9794573
IUPAC Name dicyclopentyldimethoxysilane
SMILES CO[Si](OC)(C1CCCC1)C1CCCC1

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
UN Number 3082
Formula Weight 228.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclopentyl(dimethoxy)silane
EINECSNumber (3)-4264
TSCA Yes