missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclopentanyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™
Supplier: TCI America D3381100G
Specifications
| Dicyclopentanyl Methacrylate (stabilized with MEHQ) | |
| Yellow | |
| 100 g | |
| NWAHZAIDMVNENC-UHFFFAOYSA-N | |
| 220.312 | |
| 220.31 | |
| Liquid |
| 34759-34-7 | |
| C14H20O2 | |
| Methacrylic Acid Dicyclopentanyl Ester, Methacrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Methacrylate | |
| CC(=C)C(=O)OC1CC2CC1C3C2CCC3 | |
| 13782121 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 34759-34-7 | |
| 220.312 | |
| Methacrylic Acid Dicyclopentanyl Ester, Methacrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Methacrylate | |
| CC(=C)C(=O)OC1CC2CC1C3C2CCC3 |
| C14H20O2 | |
| NWAHZAIDMVNENC-UHFFFAOYSA-N | |
| 13782121 |
Safety and Handling
EINECSNumber : (4)-1551
TSCA : No
Recommended Storage : Freezer