missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentanyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. p-7127704
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D338125G 25 g
D3381100G 100 g
D3381500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D338125G Supplier TCI America Supplier No. D338125G
Only null left

Chemical Identifiers

CAS 34759-34-7
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
InChI Key NWAHZAIDMVNENC-UHFFFAOYSA-N
Synonym Methacrylic Acid Dicyclopentanyl Ester, Methacrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Methacrylate
PubChem CID 13782121
SMILES CC(=C)C(=O)OC1CC2CC1C3C2CCC3

Specifications

Color Yellow
Quantity 25 g
Formula Weight 220.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclopentanyl Methacrylate (stabilized with MEHQ)
EINECSNumber (4)-1551
TSCA No
Recommended Storage Freezer