accessibility menu, dialog, popup

UserName

CH3CH2OO

CAS RN 34759-34-7

IUPAC Name: tricyclo[5.2.1.0²,⁶]decan-8-yl 2-methylprop-2-enoate
Synonyms: tricyclo[5.2.1.0²,⁶]decan-8-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: NWAHZAIDMVNENC-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OC1CC2CC1C1CCCC21
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results