missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl Phthalate 99.0+%, TCI America™
Supplier: TCI America P0293500G
Specifications
| Dicyclohexyl Phthalate | |
| 65°C | |
| 235°C | |
| MFCD00003849 | |
| dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
| C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 | |
| 330.424 | |
| CHEBI:34693 | |
| ≥99.0% (GC) |
| 84-61-7 | |
| White | |
| C20H26O4 | |
| 500 g | |
| VOWAEIGWURALJQ-UHFFFAOYSA-N | |
| dicyclohexyl benzene-1,2-dicarboxylate | |
| 6777 | |
| 330.42 | |
| Crystalline Powder |
Chemical Identifiers
| 84-61-7 | |
| 330.424 | |
| VOWAEIGWURALJQ-UHFFFAOYSA-N | |
| 6777 | |
| dicyclohexyl benzene-1,2-dicarboxylate |
| C20H26O4 | |
| MFCD00003849 | |
| dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
| CHEBI:34693 | |
| C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 |
Safety and Handling
EINECSNumber : (3)-1311
RTECSNumber : TI0889000
TSCA : Yes