missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl Phthalate 99.0+%, TCI America™
$57.75 - $115.51
Chemical Identifiers
| CAS | 84-61-7 |
|---|---|
| Molecular Formula | C20H26O4 |
| Molecular Weight (g/mol) | 330.424 |
| MDL Number | MFCD00003849 |
| InChI Key | VOWAEIGWURALJQ-UHFFFAOYSA-N |
| Synonym | dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 |
| PubChem CID | 6777 |
| ChEBI | CHEBI:34693 |
| IUPAC Name | dicyclohexyl benzene-1,2-dicarboxylate |
| SMILES | C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 |
Chemical Identifiers
| 84-61-7 | |
| 330.424 | |
| VOWAEIGWURALJQ-UHFFFAOYSA-N | |
| 6777 | |
| dicyclohexyl benzene-1,2-dicarboxylate |
| C20H26O4 | |
| MFCD00003849 | |
| dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
| CHEBI:34693 | |
| C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 |
Specifications
| 84-61-7 | |
| White | |
| C20H26O4 | |
| 25 g | |
| VOWAEIGWURALJQ-UHFFFAOYSA-N | |
| dicyclohexyl benzene-1,2-dicarboxylate | |
| 6777 | |
| 330.42 | |
| Crystalline Powder |
| 65°C | |
| 235°C | |
| MFCD00003849 | |
| dicyclohexyl phthalate, ergoplast fdc, unimoll 66, ergoplast.fdc, howflex cp, phthalic acid, dicyclohexyl ester, phthalic acid dicyclohexyl ester, 1,2-benzenedicarboxylic acid, dicyclohexyl ester, unii-cgd15m7h2n, ccris 6190 | |
| C1CCC(CC1)OC(=O)C2=CC=CC=C2C(=O)OC3CCCCC3 | |
| 330.424 | |
| CHEBI:34693 | |
| ≥99.0% (GC) | |
| Dicyclohexyl Phthalate |
Safety and Handling
EINECSNumber : (3)-1311
RTECSNumber : TI0889000
TSCA : Yes