missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine 97.0+%, TCI America™

Catalog No. p-7128670
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5135200MG 200 mg
D51351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5135200MG Supplier TCI America Supplier No. D5135200MG

Chemical Identifiers

CAS 303111-96-8
Molecular Formula C27H45P
Molecular Weight (g/mol) 400.63
MDL Number MFCD11656073
InChI Key DTSPXGRQYHLKLO-UHFFFAOYSA-N
Synonym dicyclohexyl 2,4,6-triisopropylphenyl phosphine, 2,4,6 tri-isopropyl phenyl di-cyclohexylphosphine, 2,4,6-tri-isopropyl phenyl di-cyclohexylphosphine, dicyclohexyl 2,4,6-tris 1-methylethyl phenyl phosphine, dicyclohexyl 2,4,6-triisopropylphenyl phosphane, acmc-1ajbl, 2,4,6 triisopropyl phenyl dicyclohexylphosphine, 1,3,5-triisopropyl-2-dicyclohexylphosphino benzene, dicyclohexyl 2,4,6-tri propan-2-yl phenyl phosphane
PubChem CID 9801140
IUPAC Name dicyclohexyl[2,4,6-tris(propan-2-yl)phenyl]phosphane
SMILES CC(C)C1=CC(C(C)C)=C(P(C2CCCCC2)C2CCCCC2)C(=C1)C(C)C

Specifications

Melting Point 139°C
Color White
Quantity 200 mg
Formula Weight 400.63
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine