accessibility menu, dialog, popup

UserName

H3CCH3H3CH3CPCH3CH3

CAS RN 303111-96-8

IUPAC Name: dicyclohexyl[2,4,6-tris(propan-2-yl)phenyl]phosphane
Synonyms: dicyclohexyl(2,4,6-triisopropylphenyl)phosphane
Molecular Weight (g/mol): 400.63
Molecular Formula: C27H45P
InChi Key: DTSPXGRQYHLKLO-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(P(C2CCCCC2)C2CCCCC2)C(=C1)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results