missing translation for 'onlineSavingsMsg'
Learn More

Dicumyl peroxide, 99%

Catalog No. p-107376
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC349881000 100 g Plastic bottle
AC349885000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC349881000 Supplier Thermo Scientific Chemicals Supplier No. 349881000
Only null left

CAS: 80-43-3 | C18H22O2 | 270.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators

Chemical Identifiers

CAS 80-43-3
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.37
MDL Number MFCD00036227
InChI Key XMNIXWIUMCBBBL-UHFFFAOYSA-N
Synonym dicumyl peroxide, cumene peroxide, cumyl peroxide, percumyl d, perkadox b, diisopropylbenzene peroxide, perkadox bc, perkadox sb, dicumenyl peroxide, peroxide, bis 1-methyl-1-phenylethyl
PubChem CID 6641
IUPAC Name 2-(2-phenylpropan-2-ylperoxy)propan-2-ylbenzene
SMILES CC(C)(C1=CC=CC=C1)OOC(C)(C)C2=CC=CC=C2

Specifications

Melting Point 38.0°C to 41.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Plastic bottle
Linear Formula [C6H5C(CH3)2]2O2
Quantity 100 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 270.37
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Dicumyl peroxide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Heating may cause a fire.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face p
EINECSNumber 201-279-3
RTECSNumber SD8150000
TSCA TSCA

RUO – Research Use Only