missing translation for 'onlineSavingsMsg'
Learn More

Diclazuril 98.0+%, TCI America™

Catalog No. D54571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54571G 1 g
D54575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54571G Supplier TCI America Supplier No. D54571G

Chemical Identifiers

CAS 101831-37-2
Molecular Formula C17H9Cl3N4O2
Molecular Weight (g/mol) 407.635
MDL Number MFCD00867203
InChI Key ZSZFUDFOPOMEET-UHFFFAOYSA-N
Synonym 2,6-Dichloro-alpha-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile
PubChem CID 456389
IUPAC Name 2-(4-chlorophenyl)-2-[2,6-dichloro-4-(3,5-dioxo-1,2,4-triazin-2-yl)phenyl]acetonitrile
SMILES C1=CC(=CC=C1C(C#N)C2=C(C=C(C=C2Cl)N3C(=O)NC(=O)C=N3)Cl)Cl

Specifications

Melting Point 297°C
Color White-Yellow
Quantity 1 g
Formula Weight 407.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diclazuril
RTECSNumber CY1697600
TSCA No