missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dickson™ 4in. (10.2cm) Recorder Replacement Charts
Supplier: Dickson™ C205
Specifications
| -22°F to +122°F | |
| 4 in. | |
| Ideal for monitoring long-term temperature trends in areas where proper temperatures are critical to product stability |
| CE, IP21 | |
| 10.16 cm | |
| 24 Hr. Chart |