missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorotris(triphenylphosphine)ruthenium(II), Premion™, 99.95% (metals basis), Ru 10.2% min
Description
Dichlorotris(triphenylphosphine)ruthenium(II) serves as a precatalyst for the hydrogenation of alkenes, nitro compounds, ketones, carboxylic acids, synthesis of furans from allenyl sulfides and C-H Activation Catalyst in a Ruthenium/Lewis Acid System
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 132°C to 134°C |
| Color | Brown to Black |
| Quantity | 1 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Very slightly soluble (with dissociation) in acetone, alcohol, chloroform, ethyl acetate, and toluene |
| Formula Weight | 958.86 |
| Percent Purity | 99.95% |
| Grade | Premion |
| Concentration or Composition (by Analyte or Components) | Ru 10.2% min. |
| Physical Form | Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 239-569-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only