missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorotitanium Diisopropoxide 95.0+%, TCI America™
$208.78 - $635.65
Chemical Identifiers
| CAS | 762-99-2 |
|---|---|
| Molecular Formula | C6H16Cl2O2Ti |
| Molecular Weight (g/mol) | 238.959 |
| MDL Number | MFCD00191416 |
| InChI Key | CGWFOWZGURYLHA-UHFFFAOYSA-L |
| Synonym | Dichloro(diisopropoxy)titanium |
| PubChem CID | 101855 |
| IUPAC Name | dichlorotitanium;propan-2-ol |
| SMILES | CC(C)O.CC(C)O.Cl[Ti]Cl |
Chemical Identifiers
| 762-99-2 | |
| 238.959 | |
| CGWFOWZGURYLHA-UHFFFAOYSA-L | |
| 101855 | |
| CC(C)O.CC(C)O.Cl[Ti]Cl |
| C6H16Cl2O2Ti | |
| MFCD00191416 | |
| Dichloro(diisopropoxy)titanium | |
| dichlorotitanium;propan-2-ol |
Specifications
| 762-99-2 | |
| C6H16Cl2O2Ti | |
| 5 g | |
| CGWFOWZGURYLHA-UHFFFAOYSA-L | |
| dichlorotitanium;propan-2-ol | |
| 101855 | |
| ≥95.0% (T) | |
| Dichlorotitanium Diisopropoxide |
| White-Yellow | |
| MFCD00191416 | |
| Dichloro(diisopropoxy)titanium | |
| CC(C)O.CC(C)O.Cl[Ti]Cl | |
| 238.959 | |
| 236.94 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer