missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorotitanium Diisopropoxide 95.0+%, TCI America™
Supplier: TCI America D225725G
Specifications
| Dichlorotitanium Diisopropoxide | |
| White-Yellow | |
| MFCD00191416 | |
| Dichloro(diisopropoxy)titanium | |
| CC(C)O.CC(C)O.Cl[Ti]Cl | |
| 238.959 | |
| 236.94 | |
| Crystalline Powder |
| 762-99-2 | |
| C6H16Cl2O2Ti | |
| 25 g | |
| CGWFOWZGURYLHA-UHFFFAOYSA-L | |
| dichlorotitanium;propan-2-ol | |
| 101855 | |
| ≥95.0% (T) |
Chemical Identifiers
| 762-99-2 | |
| 238.959 | |
| CGWFOWZGURYLHA-UHFFFAOYSA-L | |
| 101855 | |
| CC(C)O.CC(C)O.Cl[Ti]Cl |
| C6H16Cl2O2Ti | |
| MFCD00191416 | |
| Dichloro(diisopropoxy)titanium | |
| dichlorotitanium;propan-2-ol |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer