accessibility menu, dialog, popup

UserName

CAS RN 762-99-2

CH3H3COHTiOHH3CCH3ClCl

CAS RN 762-99-2

IUPAC Name: 1,1-dichloro-O'1,O1-bis(propan-2-yl)titaniumdiol
Synonyms: 1,1-dichloro-O'1,O1-diisopropyltitaniumdiol
Molecular Weight (g/mol): 238.96
Molecular Formula: C6H16Cl2O2Ti
InChi Key: CGWFOWZGURYLHA-UHFFFAOYSA-L
SMILES: Cl[Ti]Cl.CC(C)O.CC(C)O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results