missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Puriss., meets analytical spec. of Ph. Eur., NF, ≥99% (GC), Honeywell Riedel-de Haën™
Puriss., meets analytical specification of Ph. Eur., NF,≥99% (GC)
Supplier: Honeywell Chemicals 242334X25L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Dichloromethane | |
| Meets analytical specification of Ph.Eur.NF | |
| Colorless | |
| 39°C to 41°C | |
| 0.22 | |
| Glass Bottle | |
| CH2Cl2 | |
| 24.45psi (55°C), 6.83psi (20°C) | |
| 1730800 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93g/mol | |
| puriss. |
| -97°C | |
| 75-09-2 | |
| 1.318 to 1.322g/mL at 25°C, 1.325g/mL at 25°C | |
| 4 x 2.5 L | |
| 1223°F | |
| Liquid | |
| MFCD00000881 | |
| UN1593 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| ≥99% (GC) |
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Safety and Handling
P261-P281-P305 + P351 + P338
H315-H319-H335-H336-H351-H373
DOTInformation : 6.1/PGIII
EINECSNumber : 200-838-9
RTECSNumber : PA8050000