missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Puriss., meets analytical spec. of Ph. Eur., NF, ≥99% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120375
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
35 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
250 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Stainless Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045582 1 L Glass Bottle
60045583 2.5 L Glass Bottle
60045588 5 L Aluminum Bottle
60045589 6 x 1 L Glass Bottle
60045586 4 x 2.5 L Glass Bottle
60045587 4 x 5 L Aluminum Bottle
60045585 35 kg Stainless Steel Can
60045584 250 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 60045582 Supplier Honeywell Chemicals Supplier No. 242331L
Only null left

Puriss., meets analytical specification of Ph. Eur., NF,≥99% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Density 1.318 to 1.322g/mL at 25°C, 1.325g/mL at 25°C
Quantity 1 L
Explosion Limit 0.22
Ignition Point 1223°F
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 24.45psi (55°C), 6.83psi (20°C)
UN Number UN1593
Beilstein 1730800
Boiling Point 39°C to 41°C
Formula Weight 84.93g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Dichloromethane, Meets analytical specification of Ph.Eur.NF
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
Precautionary Statement H315-H319-H335-H336-H351-H373
DOTInformation 6.1/PGIII
EINECSNumber 200-838-9
RTECSNumber PA8050000