missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Purified (Methylene Chloride), Reagents
Dichloromethane, Purified (Methylene Chloride), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2195900200E
Specifications
| Dichloromethane, Purified (Methylene Chloride) | |
| 39.8°C | |
| Colorless | |
| -96.7°C | |
| CH2Cl2 | |
| 200 L | |
| Methylene Chloride | |
| ClCCl | |
| 84.93 | |
| Liquid |
| 75-09-2 | |
| -96.7°C | |
| 39.8°C | |
| Steel Drum | |
| MFCD00000881 | |
| 1.3266 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| Reagent |
Chemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| Methylene Chloride | |
| ClCCl |