missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Purified (Methylene Chloride), Reagents

Catalog No. p-200021124
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337290 1 L
01337289 200 L
01337288 20 L
01337287 4 L
01337286 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337290 Supplier Labchem Inc Supplier No. C21959001C
Only null left

Dichloromethane, Purified (Methylene Chloride), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Methylene Chloride
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Boiling Range 39.8°C
Melting Point -96.7°C
Color Colorless
Boiling Point 39.8°C
Melting Range -96.7°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.3266
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dichloromethane, Purified (Methylene Chloride)
Show More Show Less