missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99+%, extra pure, stabilized with ethanol

Catalog No. AC124050010
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124050010 1 L Glass Bottle
AC124050025 2.5 L Glass Bottle
AC124050050 5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC124050010 Supplier Thermo Scientific Chemicals Supplier No. 124050010

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-09-2 , 64-17-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Density 1.3250g/mL
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4230 to 1.4250
Residue after Evaporation 0.05% max.
Specific Gravity 1.325
Assay Percent Range 99+%
Linear Formula CH2Cl2
Viscosity 0.43 mPa.s (20°C)
Water 0.1% max. (K.F.)
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Boiling Point 39.0°C to 40.0°C
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Use personal protective equipment as required. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only