missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene

Catalog No. AC326760010
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326760010 1 L Glass Bottle
AC326760025 2.5 L Glass Bottle
AC326760100 10 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC326760010 Supplier Thermo Scientific Chemicals Supplier No. 326760010
Get 1 Extra Chemical When You Buy 2
View all promotions

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis

Chemical Identifiers

CAS 75-09-2 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Absorbance 0.5 max. at 235nm, 0.108 max. at 240nm, 0.009 max. at 260nm
Acidity 0.001% max. (HCl)
Appearance Clear liquid
Density 1.3250g/mL
Quantity 1 L
Packaging Glass Bottle
Residue after Evaporation 0.0005% max.
Specific Gravity 1.325
Assay Percent Range 99.9%
Linear Formula CH2Cl2
Viscosity 0.43 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Boiling Point 39.0°C to 40.0°C
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.9%
Grade Residue Analysis
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only