Learn More
Dichlorobis(triphenylphosphine)nickel(II), 98%
CAS: 14264-16-5 | C36H30Cl2NiP2 | 654.18 g/mol
$99.90 - $319.52
Chemical Identifiers
| CAS | 14264-16-5 |
|---|---|
| Molecular Formula | C36H30Cl2NiP2 |
| Molecular Weight (g/mol) | 654.18 |
| MDL Number | MFCD00009592 MFCD00003043 MFCD20489348 |
| InChI Key | VHQPRJDILRYZBG-UHFFFAOYSA-L |
| Synonym | bis triphenylphosphine dichloronickel ii |
| PubChem CID | 131664342 |
| IUPAC Name | nickel(2+);triphenylphosphane;dihydrochloride |
| SMILES | Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Description
Dichlorobis(triphenylphosphine)nickel(II) is used as a catalyst for cross-coupling of Grignard reagents, hydrosilylations, hydrogenation and polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
Chemical Identifiers
| 14264-16-5 | |
| 654.18 | |
| VHQPRJDILRYZBG-UHFFFAOYSA-L | |
| 131664342 | |
| Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C36H30Cl2NiP2 | |
| MFCD00009592 MFCD00003043 MFCD20489348 | |
| bis triphenylphosphine dichloronickel ii | |
| nickel(2+);triphenylphosphane;dihydrochloride |
Specifications
| 14264-16-5 | |
| Black to Gray | |
| C36H30Cl2NiP2 | |
| 10 g | |
| bis triphenylphosphine dichloronickel ii | |
| VHQPRJDILRYZBG-UHFFFAOYSA-L | |
| nickel(2+);triphenylphosphane;dihydrochloride | |
| 131664342 | |
| 98% | |
| Dichlorobis(triphenylphosphine)nickel(II) |
| ∼225°C (decomposition) | |
| Odorless | |
| MFCD00009592 MFCD00003043 MFCD20489348 | |
| Moisture sensitive | |
| Insoluble in water. | |
| Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 654.18 | |
| 654.18 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H312+H332-H314-H317-H335-H350
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 238-154-8
RTECSNumber : QR6170000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only